Assay Details
| Gene Symbol: SOS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085208.2 | NP_001078677.1 | 5 - 6 | 593 | 119 | |
| NM_001344222.1 | NP_001331291.1 | 4 - 5 | 685 | 119 | ||
| NM_123144.4 | NP_198601.2 | 5 - 6 | 560 | 119 | ||
| GenBank mRNA | AF404865.2 | - | 4 - 5 | 364 | 119 | |
| AY084419.1 | - | 4 - 5 | 394 | 119 | ||
| AY136333.1 | - | 5 - 6 | 452 | 119 | ||
| BT008815.1 | - | 4 - 5 | 364 | 119 | ||
More Information
Set Membership: |
|

SOS4
Genomic Map