Assay Details
| Gene Symbol: AT5G38160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123176.3 | NP_198632.1 | 1 | 333 | 113 | |
| GenBank mRNA | AK118130.1 | - | 1 | 338 | 113 | |
| BT005612.1 | - | 1 | 264 | 113 | ||
More Information
Set Membership: |
|

AT5G38160
Genomic Map