Assay Details
| Gene Symbol: AT5G38195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123180.1 | NP_568555.1 | 1 | 162 | 87 | |
| GenBank mRNA | BT024655.1 | - | 1 | 81 | 87 | |
| DQ447010.1 | - | 1 | 81 | 87 | ||
More Information
Set Membership: |
|

AT5G38195
Genomic Map