Assay Details
| Gene Symbol: SYP131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337487.1 | NP_001319461.1 | 12 - 13 | 920 | 143 | |
| NM_001337488.1 | NP_001326903.1 | 11 - 12 | 825 | 143 | ||
More Information
Set Membership: |
|

SYP131
Genomic Map