Assay Details
| Gene Symbol: SAUR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111255.4 | NP_187034.1 | 1 | 527 | 177 | |
| GenBank mRNA | AK117273.1 | - | 1 | 406 | 177 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAUR27| Gene Symbol: SAUR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111255.4 | NP_187034.1 | 1 | 527 | 177 | |
| GenBank mRNA | AK117273.1 | - | 1 | 406 | 177 | |
Set Membership: |
|