Assay Details
| Gene Symbol: GDH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337494.1 | NP_001326159.1 | 1 - 2 | 182 | 111 | |
| NM_111262.2 | NP_187041.1 | 1 - 2 | 243 | 111 | ||
More Information
Set Membership: |
|

GDH3
Genomic Map