Assay Details
| Gene Symbol: AT3G03920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111263.4 | NP_566217.1 | 2 - 3 | 512 | 111 | |
| GenBank mRNA | AY063874.1 | - | 2 - 3 | 409 | 111 | |
| AY091234.1 | - | 2 - 3 | 318 | 111 | ||
More Information
Set Membership: |
|

AT3G03920
Genomic Map