Assay Details
| Gene Symbol: AT3G03920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111263.4 | NP_566217.1 | 3 - 4 | 641 | 88 | |
| GenBank mRNA | AY063874.1 | - | 3 - 4 | 538 | 88 | |
| AY091234.1 | - | 3 - 4 | 447 | 88 | ||
More Information
Set Membership: |
|

AT3G03920
Genomic Map