Assay Details
| Gene Symbol: NAC047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125096.1 | NP_001118568.1 | 1 - 2 | 296 | 73 | |
| NM_111278.4 | NP_187057.2 | 1 - 2 | 427 | 73 | ||
| GenBank mRNA | AK227417.1 | - | 1 - 2 | 287 | 73 | |
| BT005985.1 | - | 1 - 2 | 169 | 73 | ||
More Information
Set Membership: |
|

NAC047
Genomic Map