Assay Details
| Gene Symbol: NAC047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111278.4 | NP_187057.2 | 2 - 3 | 775 | 88 | |
| GenBank mRNA | AK227417.1 | - | 2 - 3 | 635 | 88 | |
| BT005985.1 | - | 2 - 3 | 517 | 88 | ||
More Information
Set Membership: |
|

NAC047
Genomic Map