Assay Details
| Gene Symbol: ARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114070.4 | NP_566861.1 | 1 - 2 | 989 | 72 | |
| GenBank mRNA | AY084321.1 | - | 1 - 2 | 950 | 72 | |
| AY150426.1 | - | 1 - 2 | 877 | 72 | ||
More Information
Set Membership: |
|

ARP6
Genomic Map