Assay Details
| Gene Symbol: AT3G42050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114072.4 | NP_189791.1 | 7 - 8 | 901 | 109 | |
| GenBank mRNA | AY091685.1 | - | 6 - 7 | 482 | 109 | |
More Information
Set Membership: |
|

AT3G42050
Genomic Map