Assay Details
| Gene Symbol: LTPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114240.4 | NP_189958.1 | 2 - 3 | 654 | 62 | |
| GenBank mRNA | AB246324.1 | - | 2 - 3 | 444 | 62 | |
| AY045908.1 | - | 2 - 3 | 470 | 62 | ||
| AY079420.1 | - | 2 - 3 | 444 | 62 | ||
More Information
Set Membership: |
|

LTPG2
Genomic Map