Assay Details
| Gene Symbol: AT3G44050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114273.4 | NP_189991.2 | 11 - 12 | 1465 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G44050| Gene Symbol: AT3G44050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114273.4 | NP_189991.2 | 11 - 12 | 1465 | 118 | |
Set Membership: |
|