Assay Details
| Gene Symbol: CAF1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114294.3 | NP_190012.1 | 1 | 1111 | 165 | |
| GenBank mRNA | AY050848.1 | - | 1 | 849 | 166 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAF1a| Gene Symbol: CAF1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114294.3 | NP_190012.1 | 1 | 1111 | 165 | |
| GenBank mRNA | AY050848.1 | - | 1 | 849 | 166 | |
Set Membership: |
|