Assay Details
| Gene Symbol: HIRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339149.1 | NP_001319681.1 | 5 - 6 | 2551 | 60 | |
| NM_001339150.1 | NP_001326052.1 | 5 - 6 | 2472 | 60 | ||
| NM_001339151.1 | NP_001326053.1 | 4 - 5 | 2555 | 60 | ||
| NM_114321.3 | NP_190039.2 | 5 - 6 | 2367 | 60 | ||
More Information
Set Membership: |
|

HIRA
Genomic Map