Assay Details
| Gene Symbol: HDA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114336.4 | NP_190054.2 | 8 - 9 | 1086 | 103 | |
| GenBank mRNA | BT002003.1 | - | 8 - 9 | 996 | 103 | |
| BT006576.1 | - | 8 - 9 | 829 | 103 | ||
More Information
Set Membership: |
|

HDA9
Genomic Map