Assay Details
| Gene Symbol: RPL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114358.4 | NP_190075.1 | 2 - 3 | 388 | 68 | |
| GenBank mRNA | BT000914.1 | - | 2 - 3 | 278 | 68 | |
| Z11129.1 | - | 287 | 68 | |||
More Information
Set Membership: |
|

RPL9
Genomic Map