Assay Details
| Gene Symbol: AT3G45040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339189.1 | NP_001325870.1 | 3 - 4 | 594 | 110 | |
| NM_114373.4 | NP_190090.2 | 3 - 4 | 594 | 110 | ||
More Information
Set Membership: |
|

AT3G45040
Genomic Map