Assay Details
| Gene Symbol: AT3G45040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339189.1 | NP_001325870.1 | 7 - 8 | 1067 | 115 | |
| NM_001339190.1 | NP_001325871.1 | 7 - 8 | 1064 | 115 | ||
| NM_114373.4 | NP_190090.2 | 7 - 8 | 1067 | 115 | ||
More Information
Set Membership: |
|

AT3G45040
Genomic Map