Assay Details
| Gene Symbol: LOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114383.3 | NP_566875.1 | 1 - 2 | 547 | 65 | |
| GenBank mRNA | L23968.1 | - | 1 - 2 | 380 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOX2| Gene Symbol: LOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114383.3 | NP_566875.1 | 1 - 2 | 547 | 65 | |
| GenBank mRNA | L23968.1 | - | 1 - 2 | 380 | 65 | |
Set Membership: |
|