Assay Details
| Gene Symbol: MLO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114398.2 | NP_566879.1 | 2 - 3 | 393 | 139 | |
| GenBank mRNA | AF369564.1 | - | 2 - 3 | 247 | 139 | |
More Information
Set Membership: |
|

MLO3
Genomic Map