Assay Details
| Gene Symbol: AT4G23760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118507.3 | NP_194107.2 | 1 - 2 | 282 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G23760| Gene Symbol: AT4G23760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118507.3 | NP_194107.2 | 1 - 2 | 282 | 109 | |
Set Membership: |
|