Assay Details
| Gene Symbol: ZIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126440.3 | NP_178488.1 | 1 - 2 | 750 | 103 | |
| GenBank mRNA | BT015108.1 | - | 1 - 2 | 638 | 103 | |
More Information
Set Membership: |
|

ZIP7
Genomic Map