Assay Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126484.5 | NP_178532.2 | 5 - 6 | 949 | 66 | |
| GenBank mRNA | AY099564.1 | - | 5 - 6 | 853 | 66 | |
| BT001247.1 | - | 5 - 6 | 835 | 66 | ||
More Information
Set Membership: |
|

CPZ
Genomic Map