Assay Details
| Gene Symbol: AT2G05420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126565.2 | NP_178610.1 | 1 - 2 | 112 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G05420| Gene Symbol: AT2G05420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126565.2 | NP_178610.1 | 1 - 2 | 112 | 70 | |
Set Membership: |
|