Assay Details
| Gene Symbol: GRP-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036258.2 | NP_001031335.1 | 1 - 2 | 309 | 87 | |
| NM_001036259.1 | NP_001031336.1 | 1 - 2 | 138 | 87 | ||
| NM_001036260.1 | NP_001031337.1 | 1 - 2 | 138 | 87 | ||
| NM_001036261.1 | NP_001031338.1 | 1 - 2 | 138 | 87 | ||
| NM_001124805.1 | NP_001118277.1 | 1 - 2 | 138 | 87 | ||
| NM_126575.4 | NP_178620.1 | 1 - 2 | 309 | 87 | ||
| GenBank mRNA | AY065139.1 | - | 1 - 2 | 138 | 87 | |
| AY081567.1 | - | 1 - 2 | 83 | 87 | ||
| BT000704.1 | - | 1 - 2 | 136 | 87 | ||
More Information
Set Membership: |
|

GRP-3
Genomic Map