Assay Details
| Gene Symbol: PAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036263.1 | NP_001031340.1 | 3 - 4 | 391 | 89 | |
| NM_001335288.1 | NP_001324458.1 | 3 - 4 | 423 | 89 | ||
| NM_126597.4 | NP_178641.1 | 3 - 4 | 464 | 89 | ||
| GenBank mRNA | AF043519.1 | - | 3 - 4 | 373 | 89 | |
| AY052679.1 | - | 3 - 4 | 316 | 89 | ||
| AY143829.1 | - | 3 - 4 | 257 | 89 | ||
More Information
Set Membership: |
|

PAA2
Genomic Map