Assay Details
| Gene Symbol: PAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126597.4 | NP_178641.1 | 7 - 8 | 799 | 100 | |
| GenBank mRNA | AF043519.1 | - | 7 - 8 | 708 | 100 | |
| AY052679.1 | - | 7 - 8 | 651 | 100 | ||
| AY143829.1 | - | 7 - 8 | 592 | 100 | ||
More Information
Set Membership: |
|

PAA2
Genomic Map