Assay Details
| Gene Symbol: RIPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126607.3 | NP_178651.1 | 1 - 2 | 772 | 93 | |
| GenBank mRNA | AY056245.1 | - | 1 - 2 | 445 | 93 | |
| AY113963.1 | - | 1 - 2 | 406 | 93 | ||
More Information
Set Membership: |
|

RIPK
Genomic Map