Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335289.1 | NP_001318204.1 | 11 - 12 | 1060 | 62 | |
| NM_001335290.1 | NP_001324184.1 | 10 - 11 | 880 | 62 | ||
| NM_001335291.1 | NP_001324181.1 | 11 - 12 | 929 | 62 | ||
| NM_001335292.1 | NP_001324182.1 | 11 - 12 | 925 | 62 | ||
| NM_001335293.1 | NP_001324183.1 | 11 - 12 | 912 | 62 | ||
| NM_126614.3 | NP_565332.1 | 11 - 12 | 934 | 62 | ||
| GenBank mRNA | AY086482.1 | - | 11 - 12 | 834 | 62 | |
More Information
Set Membership: |
|

FIP1
Genomic Map