Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335289.1 | NP_001318204.1 | 12 - 13 | 1157 | 66 | |
| NM_001335290.1 | NP_001324184.1 | 11 - 12 | 977 | 66 | ||
| NM_126614.3 | NP_565332.1 | 12 - 13 | 1031 | 66 | ||
| GenBank mRNA | AY086482.1 | - | 12 - 13 | 931 | 66 | |
More Information
Set Membership: |
|

FIP1
Genomic Map