Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335289.1 | NP_001318204.1 | 6 - 7 | 706 | 71 | |
| NM_001335290.1 | NP_001324184.1 | 5 - 6 | 526 | 71 | ||
| NM_001335291.1 | NP_001324181.1 | 6 - 7 | 575 | 71 | ||
| NM_001335292.1 | NP_001324182.1 | 6 - 7 | 571 | 71 | ||
| NM_001335293.1 | NP_001324183.1 | 6 - 7 | 558 | 71 | ||
| NM_126614.3 | NP_565332.1 | 6 - 7 | 580 | 71 | ||
| GenBank mRNA | AY086482.1 | - | 6 - 7 | 480 | 71 | |
More Information
Set Membership: |
|

FIP1
Genomic Map