Assay Details
| Gene Symbol: FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335289.1 | NP_001318204.1 | 8 - 9 | 860 | 118 | |
| NM_001335290.1 | NP_001324184.1 | 7 - 8 | 680 | 118 | ||
| NM_001335291.1 | NP_001324181.1 | 8 - 9 | 729 | 118 | ||
| NM_001335292.1 | NP_001324182.1 | 8 - 9 | 725 | 118 | ||
| NM_001335293.1 | NP_001324183.1 | 8 - 9 | 712 | 118 | ||
| NM_126614.3 | NP_565332.1 | 8 - 9 | 734 | 118 | ||
| GenBank mRNA | AY086482.1 | - | 8 - 9 | 634 | 118 | |
More Information
Set Membership: |
|

FIP1
Genomic Map