Assay Details
| Gene Symbol: AT2G07170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126689.4 | NP_178730.2 | 2 - 3 | 837 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G07170| Gene Symbol: AT2G07170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126689.4 | NP_178730.2 | 2 - 3 | 837 | 71 | |
Set Membership: |
|