Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 20 - 21 | 2126 | 119 | |
| NM_001335346.1 | NP_001323940.1 | 20 - 21 | 2126 | 119 | ||
| NM_001335347.1 | NP_001323936.1 | 19 - 20 | 2043 | 119 | ||
| NM_001335348.1 | NP_001323937.1 | 19 - 20 | 1972 | 119 | ||
| NM_001335349.1 | NP_001323938.1 | 17 - 18 | 1903 | 119 | ||
| NM_126770.8 | NP_178811.7 | 19 - 20 | 1983 | 119 | ||
More Information
Set Membership: |
|

ABCC13
Genomic Map