Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 11 - 12 | 1145 | 78 | |
| NM_001335346.1 | NP_001323940.1 | 11 - 12 | 1145 | 78 | ||
| NM_001335347.1 | NP_001323936.1 | 10 - 11 | 1062 | 78 | ||
| NM_001335348.1 | NP_001323937.1 | 10 - 11 | 991 | 78 | ||
| NM_001335349.1 | NP_001323938.1 | 8 - 9 | 922 | 78 | ||
| NM_126770.8 | NP_178811.7 | 10 - 11 | 1002 | 78 | ||
More Information
Set Membership: |
|

ABCC13
Genomic Map