Assay Details
| Gene Symbol: MAK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124823.2 | NP_001118295.1 | 7 - 8 | 1175 | 100 | |
| NM_001335364.1 | NP_001324918.1 | 7 - 8 | 1150 | 100 | ||
| NM_001335365.1 | NP_001324917.1 | 7 - 8 | 1069 | 100 | ||
| NM_126831.4 | NP_178872.2 | 7 - 8 | 1232 | 100 | ||
More Information
Set Membership: |
|

MAK10
Genomic Map