Assay Details
| Gene Symbol: ABH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126934.4 | NP_565356.1 | 11 - 12 | 1231 | 117 | |
| GenBank mRNA | AF268377.1 | - | 11 - 12 | 1231 | 117 | |
| AF272891.1 | - | 11 - 12 | 1098 | 117 | ||
More Information
Set Membership: |
|

ABH1
Genomic Map