Assay Details
| Gene Symbol: ABH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126934.4 | NP_565356.1 | 6 - 7 | 682 | 144 | |
| GenBank mRNA | AF268377.1 | - | 6 - 7 | 682 | 144 | |
| AF272891.1 | - | 6 - 7 | 549 | 144 | ||
More Information
Set Membership: |
|

ABH1
Genomic Map