Assay Details
| Gene Symbol: AT2G13720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126949.1 | NP_178993.1 | 3 - 4 | 325 | 163 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G13720| Gene Symbol: AT2G13720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126949.1 | NP_178993.1 | 3 - 4 | 325 | 163 | |
Set Membership: |
|