Assay Details
| Gene Symbol: SERK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335399.1 | NP_001318220.1 | 3 - 4 | 328 | 103 | |
| NM_126956.4 | NP_179000.3 | 2 - 3 | 398 | 103 | ||
| GenBank mRNA | AY094412.1 | - | 2 - 3 | 214 | 103 | |
| FJ708692.1 | - | 2 - 3 | 225 | 103 | ||
More Information
Set Membership: |
|

SERK5
Genomic Map