Assay Details
| Gene Symbol: SERK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335399.1 | NP_001318220.1 | 8 - 9 | 739 | 67 | |
| NM_001335400.1 | NP_001324219.1 | 6 - 7 | 875 | 67 | ||
| NM_001335401.1 | NP_001324220.1 | 5 - 6 | 448 | 67 | ||
| NM_126956.4 | NP_179000.3 | 7 - 8 | 809 | 67 | ||
| GenBank mRNA | AY094412.1 | - | 7 - 8 | 625 | 67 | |
| BT001116.1 | - | 4 - 5 | 285 | 67 | ||
| FJ708692.1 | - | 7 - 8 | 636 | 67 | ||
More Information
Set Membership: |
|

SERK5
Genomic Map