Assay Details
| Gene Symbol: ALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126957.2 | NP_565359.1 | 4 - 5 | 357 | 82 | |
| GenBank mRNA | AY057526.1 | - | 4 - 5 | 328 | 82 | |
| AY143898.1 | - | 4 - 5 | 285 | 82 | ||
| AY518702.1 | - | 4 - 5 | 285 | 82 | ||
More Information
Set Membership: |
|

ALD1
Genomic Map