Assay Details
| Gene Symbol: AGL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335416.1 | NP_001324071.1 | 2 - 3 | 291 | 67 | |
| NM_126990.3 | NP_179033.1 | 2 - 3 | 291 | 67 | ||
| GenBank mRNA | AK228244.1 | - | 2 - 3 | 291 | 67 | |
| BT005861.1 | - | 1 - 2 | 178 | 67 | ||
More Information
Set Membership: |
|

AGL44
Genomic Map