Assay Details
| Gene Symbol: AGL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335416.1 | NP_001324071.1 | 5 - 6 | 541 | 132 | |
| NM_126990.3 | NP_179033.1 | 5 - 6 | 541 | 132 | ||
| GenBank mRNA | AK228244.1 | - | 5 - 6 | 541 | 132 | |
| BT005861.1 | - | 4 - 5 | 428 | 132 | ||
More Information
Set Membership: |
|

AGL44
Genomic Map