Assay Details
| Gene Symbol: AGL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126990.3 | NP_179033.1 | 8 - 9 | 808 | 125 | |
| GenBank mRNA | AK228244.1 | - | 8 - 9 | 808 | 125 | |
More Information
Set Membership: |
|

AGL44
Genomic Map