Assay Details
| Gene Symbol: PIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335417.1 | NP_001325175.1 | 2 - 3 | 549 | 55 | |
| NM_001335418.1 | NP_001325176.1 | 2 - 3 | 396 | 55 | ||
| NM_126994.4 | NP_179037.2 | 2 - 3 | 396 | 55 | ||
| NM_201725.3 | NP_973454.1 | 1 - 2 | 461 | 55 | ||
| GenBank mRNA | AY059916.1 | - | 2 - 3 | 301 | 55 | |
| AY114690.1 | - | 2 - 3 | 291 | 55 | ||
More Information
Set Membership: |
|

PIP
Genomic Map