Assay Details
| Gene Symbol: PIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335417.1 | NP_001325175.1 | 6 - 7 | 922 | 77 | |
| NM_001335418.1 | NP_001325176.1 | 6 - 7 | 769 | 77 | ||
| NM_126994.4 | NP_179037.2 | 6 - 7 | 769 | 77 | ||
| NM_201725.3 | NP_973454.1 | 5 - 6 | 834 | 77 | ||
| GenBank mRNA | AY059916.1 | - | 6 - 7 | 674 | 77 | |
| AY114690.1 | - | 6 - 7 | 664 | 77 | ||
More Information
Set Membership: |
|

PIP
Genomic Map