Assay Details
| Gene Symbol: FUS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100089.3 | NP_563645.1 | 6 - 7 | 775 | 97 | |
| NM_179245.3 | NP_849576.1 | 6 - 7 | 775 | 97 | ||
| NM_202010.3 | NP_973739.1 | 6 - 7 | 775 | 97 | ||
| GenBank mRNA | AF063852.2 | - | 6 - 7 | 579 | 97 | |
| AF372934.1 | - | 6 - 7 | 610 | 97 | ||
| AF395065.1 | - | 6 - 7 | 538 | 97 | ||
| AF395066.1 | - | 6 - 7 | 538 | 97 | ||
| AY087971.1 | - | 6 - 7 | 613 | 97 | ||
| AY133518.1 | - | 6 - 7 | 535 | 97 | ||
More Information
Set Membership: |
|

FUS5
Genomic Map